The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053214.2 | NP_444444.2 | 16 - 17 | 1837 | 60 |
GenBank mRNA | AK036840.1 | - | 6 - 7 | 1670 | 60 |
AK131188.1 | - | 6 - 7 | 530 | 60 | |
AK153908.1 | - | 16 - 17 | 1800 | 60 | |
AK158333.1 | - | 16 - 17 | 1752 | 60 | |
BC046502.1 | - | 16 - 17 | 1825 | 60 | |
X97650.1 | - | 16 - 17 | 1755 | 60 |