Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fsd1
Entrez Gene ID
240121
Gene Name
fibronectin type 3 and SPRY domain-containing protein
Gene Aliases
-
Chromosome Location
Chr.17: 55986462 - 55996883 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_183178.2 NP_899001.1 7 - 8 828 98
GenBank mRNA AK140132.1 - 7 - 8 862 98
AK144257.1 - 7 - 8 763 98
BC055106.1 - 7 - 8 828 98

View Full Product Details