The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024432.2 | NP_077752.1 | 1 - 2 | 150 | 61 |
GenBank mRNA | AF272895.1 | - | 1 - 2 | 165 | 61 |
AK008942.1 | - | 1 - 2 | 160 | 61 | |
AK075705.1 | - | 1 - 2 | 179 | 61 | |
AK082510.1 | - | - | 189 | 61 | |
AK148170.1 | - | - | 118 | 61 | |
AK161273.1 | - | 1 - 2 | 171 | 61 | |
BC016412.1 | - | 1 - 2 | 79 | 61 | |
BC049963.1 | - | 1 - 2 | 150 | 61 |