Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ubxn6
Entrez Gene ID
66530
Gene Name
UBX domain protein 6
Gene Aliases
1200008L11Rik, 2210415J11Rik, AU040909, Ubxd1, Ubxdc2
Chromosome Location
Chr.17: 56068249 - 56074989 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024432.2 NP_077752.1 2 - 3 315 62
GenBank mRNA AF272895.1 - 2 - 3 330 62
AK008942.1 - 2 - 3 324 62
AK009117.1 - 1 - 2 97 62
AK075705.1 - 2 - 3 343 62
AK082510.1 - - 354 62
AK148170.1 - - 283 62
AK161273.1 - 2 - 3 336 62
BC016412.1 - 2 - 3 244 62
BC049963.1 - 2 - 3 315 62

View Full Product Details