Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
H2-T23
Entrez Gene ID
15040
Gene Name
histocompatibility 2, T region locus 23
Gene Aliases
37b, 37c, H-2T23, H2-Qa1, Qa-1, Qa-1(b), Qa1, Qed-1, T18c, T18c(37), T23b, T23d
Chromosome Location
Chr.17: 36029765 - 36032850 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010398.3 NP_034528.1 2 - 3 369 65
GenBank mRNA AB359230.2 - 2 - 3 369 65
AF057279.2 - - 337 65
AF100700.1 - - 316 65
AF100701.1 - - 316 65
AF100702.1 - 2 - 3 316 65
AK133502.1 - 2 - 3 369 65
BC099472.1 - 2 - 3 358 65
M11284.1 - - 370 65

View Full Product Details