The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163764.1 | NP_001157236.1 | 1 - 2 | 68 | 63 |
GenBank mRNA | CX220762.1 | - | 1 - 2 | 31 | 63 |