Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rab6a
Entrez Gene ID
19346
Gene Name
RAB6A, member RAS oncogene family
Gene Aliases
2610028L11Rik, AA419671, Rab6
Chromosome Location
Chr.7: 100607586 - 100641268 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163663.1 NP_001157135.1 3 - 4 823 100
GenBank mRNA AK154012.1 - 3 - 4 720 100
BC019118.1 - 3 - 4 455 100
BE134038.1 - 3 - 4 402 100

View Full Product Details