Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gp2
Entrez Gene ID
67133
Gene Name
glycoprotein 2 (zymogen granule membrane)
Gene Aliases
2310037I18Rik, AV060639
Chromosome Location
Chr.7: 119442542 - 119459272 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025989.3 NP_080265.2 7 - 8 1388 74
GenBank mRNA AK009323.1 - 7 - 8 1389 74
AK009466.1 - 2 - 3 285 74
AK009558.1 - 1 - 2 98 74
AK009661.1 - 7 - 8 1389 74
AK009843.1 - 7 - 8 1342 74
BC119573.2 - 7 - 8 1383 74

View Full Product Details