Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Hras
- Entrez Gene ID
-
15461
- Gene Name
- Harvey rat sarcoma virus oncogene
- Gene Aliases
-
H-ras, Ha-ras, Harvey-ras, Hras-1, Hras1, Kras2, c-H-ras, c-Ha-ras, c-rasHa, ras
- Chromosome Location
-
Chr.7: 141189934 - 141194004 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC011083.1 |
- |
3 - 4 |
375 |
76 |
View Full Product Details