The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001111049.1 | NP_001104519.1 | 5 - 6 | 596 | 67 |
NM_001111050.1 | NP_001104520.1 | 4 - 5 | 471 | 67 | |
NM_009842.3 | NP_033972.2 | 5 - 6 | 714 | 67 | |
GenBank mRNA | BC012236.1 | - | 4 - 5 | 393 | 67 |
BQ896459.1 | - | 5 - 6 | 732 | 67 | |
D89290.1 | - | 5 - 6 | 683 | 67 | |
DV063036.1 | - | 5 - 6 | 623 | 67 | |
U89772.1 | - | 4 - 5 | 438 | 67 |