Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prss36
Entrez Gene ID
77613
Gene Name
protease, serine 36
Gene Aliases
C330007D15Rik
Chromosome Location
Chr.7: 127932638 - 127946725 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK021185.1 - 2 - 3 150 61

View Full Product Details