The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177897.3 | NP_808565.2 | 14 - 15 | 2189 | 61 |
GenBank mRNA | AB114827.1 | - | 14 - 15 | 2189 | 61 |
BC062653.1 | - | 10 - 11 | 1879 | 61 | |
BC079533.1 | - | 3 - 4 | 1092 | 61 |