The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024134.2 | NP_001019305.1 | 4 - 5 | 803 | 73 |
NM_001177872.1 | NP_001171343.1 | 4 - 5 | 826 | 73 | |
GenBank mRNA | AK007445.1 | - | 4 - 5 | 827 | 73 |
BC027186.1 | - | 4 - 5 | 803 | 73 |