The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033405.2 | NP_001028577.2 | 1 - 2 | 555 | 101 |
GenBank mRNA | AK013294.1 | - | 1 - 2 | 239 | 101 |
AK037593.1 | - | 1 - 2 | 207 | 101 | |
AK132018.1 | - | 2 - 3 | 587 | 101 | |
DQ087184.1 | - | 1 - 2 | 79 | 101 | |
DQ341272.1 | - | 1 - 2 | 654 | 101 |