The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027225.1 | NP_081501.1 | 12 - 13 | 3718 | 66 |
NM_177025.5 | NP_795999.2 | 13 - 14 | 3829 | 66 | |
GenBank mRNA | AK037010.1 | - | 14 - 15 | 3823 | 66 |
AK052533.1 | - | 12 - 13 | 3667 | 66 | |
AK135859.1 | - | 4 - 5 | 2057 | 66 | |
AK144943.1 | - | 14 - 15 | 3876 | 66 | |
AK173076.1 | - | 12 - 13 | 3549 | 66 | |
AY308746.1 | - | 13 - 14 | 3829 | 66 | |
BC067007.1 | - | 12 - 13 | 3717 | 66 |