Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cerkl
Entrez Gene ID
228094
Gene Name
ceramide kinase-like
Gene Aliases
Gm1958, Rp26
Chromosome Location
Chr.2: 79331105 - 79430520 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001048176.1 NP_001041641.1 9 - 10 1208 121
GenBank mRNA BC046474.1 - 9 - 10 1314 121
CJ235603.1 - 3 - 4 139 121
DT912627.1 - 5 - 6 404 121
HQ426680.1 - 9 - 10 1208 121
HQ426681.1 - 7 - 8 1001 121
HQ426682.1 - 6 - 7 869 121
HQ426683.1 - 5 - 6 626 121
HQ426684.1 - 8 - 9 1076 121
HQ426686.1 - 8 - 9 1065 121
HQ426687.1 - 9 - 10 1339 121
HQ426688.1 - 9 - 10 1355 121
HQ426689.1 - 8 - 9 1223 121
HQ426690.1 - 9 - 10 1425 121
HQ426691.1 - - 1007 121
HQ426692.1 - - 1138 121
HQ426693.1 - - 1048 121
HQ426694.1 - - 961 121
HQ426695.1 - - 1180 121
HQ426698.1 - - 973 121
HQ426699.1 - - 1037 121
HQ426700.1 - - 1311 121
HQ426701.1 - - 1327 121
HQ426702.1 - 8 - 9 790 121
HQ426703.1 - 6 - 7 583 121
HQ426704.1 - 8 - 9 790 121
HQ426705.1 - 7 - 8 647 121
HQ426706.1 - 7 - 8 734 121
HQ426707.1 - 7 - 8 881 121
HQ426709.1 - 5 - 6 482 121
HQ426711.1 - 5 - 6 629 121

View Full Product Details