The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145824.1 | NP_001139296.1 | 15 - 16 | 3643 | 60 |
NM_010434.2 | NP_034564.2 | 15 - 16 | 3646 | 60 | |
GenBank mRNA | AF077660.1 | - | 15 - 16 | 3400 | 60 |
AF170305.1 | - | 14 - 15 | 3110 | 60 | |
AF305238.1 | - | 14 - 15 | 3108 | 60 | |
AK140130.1 | - | 15 - 16 | 3749 | 60 | |
BC131663.1 | - | 15 - 16 | 3278 | 60 | |
BC131664.1 | - | 15 - 16 | 3274 | 60 |