Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Plin5
Entrez Gene ID
66968
Gene Name
perilipin 5
Gene Aliases
2310076L09Rik, AI415325, AW109675, Lsdp5, MLDP, PAT-1
Chromosome Location
Chr.17: 56111597 - 56117606 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077348.1 NP_001070816.1 6 - 7 934 55
NM_025874.3 NP_080150.2 7 - 8 880 55
GenBank mRNA AK010202.1 - 7 - 8 880 55
AK075872.1 - 7 - 8 877 55
AY919875.1 - 6 - 7 893 55
BC024138.1 - 7 - 8 860 55
DQ473305.1 - 7 - 8 860 55

View Full Product Details