Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prss36
Entrez Gene ID
77613
Gene Name
protease, serine 36
Gene Aliases
C330007D15Rik
Chromosome Location
Chr.7: 127932638 - 127946725 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081374.1 NP_001074843.2 4 - 5 555 71

View Full Product Details