Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ric8
Entrez Gene ID
101489
Gene Name
resistance to inhibitors of cholinesterase 8 homolog (C. elegans)
Gene Aliases
AI114950, RIC-8, Ric8a
Chromosome Location
Chr.7: 140857397 - 140863731 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053194.4 NP_444424.1 7 - 8 1286 67
GenBank mRNA AF288813.1 - 7 - 8 1285 67
AK031466.1 - 7 - 8 1233 67
AK080192.1 - 7 - 8 1229 67
AK155282.1 - 7 - 8 1261 67
AK165536.1 - 7 - 8 1263 67
AK167746.1 - 7 - 8 1289 67
AK169367.1 - 7 - 8 1250 67
BC022917.1 - 7 - 8 1263 67

View Full Product Details