Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rrm1
Entrez Gene ID
20133
Gene Name
ribonucleotide reductase M1
Gene Aliases
RnrM1
Chromosome Location
Chr.7: 102441695 - 102469771 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009103.3 NP_033129.2 13 - 14 1804 105
GenBank mRNA AK082135.1 - 13 - 14 1682 105
AK137075.1 - 13 - 14 1637 105
AK168586.1 - 13 - 14 1671 105
AK210911.1 - 1 - 2 96 105
AK215988.1 - 1 - 2 96 105
BC003299.1 - 13 - 14 1672 105
BC016450.1 - 13 - 14 1632 105
BC066217.1 - 13 - 14 1682 105

View Full Product Details