Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Smoc2
Entrez Gene ID
64074
Gene Name
SPARC related modular calcium binding 2
Gene Aliases
1700056C05Rik, 5430426J21Rik, Smoc2l
Chromosome Location
Chr.17: 14279487 - 14404790 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022315.2 NP_071710.2 12 - 13 1532 69
GenBank mRNA AK030958.1 - 12 - 13 1501 69

View Full Product Details