Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrfn2
Entrez Gene ID
70530
Gene Name
leucine rich repeat and fibronectin type III domain containing 2
Gene Aliases
5730420O05Rik, SALM1, mKIAA1246
Chromosome Location
Chr.17: 48931566 - 49097590 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027452.3 NP_081728.2 2 - 3 1664 65
GenBank mRNA AK017594.1 - 2 - 3 1868 65
AK044375.1 - 2 - 3 1541 65
AK122476.1 - 2 - 3 1619 65
BC139418.1 - 2 - 3 1664 65

View Full Product Details