Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Smn1
Entrez Gene ID
20595
Gene Name
survival motor neuron 1
Gene Aliases
AI849087, Gemin1, Smn
Chromosome Location
Chr.13: 100123205 - 100137699 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252629.1 NP_001239558.1 3 - 4 366 69
NM_011420.2 NP_035550.1 3 - 4 302 69
GenBank mRNA AK144778.1 - 3 - 4 303 69
AK167832.1 - 3 - 4 300 69
BC045158.1 - 3 - 4 266 69
EU234021.1 - 3 - 4 366 69
U63294.1 - 3 - 4 280 69
U77714.1 - 3 - 4 263 69
Y12835.1 - 3 - 4 288 69

View Full Product Details