The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030026.2 | NP_084302.1 | 12 - 13 | 1273 | 134 |
GenBank mRNA | AK132265.1 | - | 12 - 13 | 1253 | 134 |
AK143233.1 | - | 12 - 13 | 1274 | 134 | |
AK145564.1 | - | 12 - 13 | 1225 | 134 | |
AK146844.1 | - | 12 - 13 | 1233 | 134 | |
AK146865.1 | - | 12 - 13 | 1223 | 134 | |
BC058987.1 | - | 4 - 5 | 287 | 134 | |
BC141193.1 | - | 12 - 13 | 1239 | 134 | |
BC141194.1 | - | 12 - 13 | 1239 | 134 |