Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mccc2
Entrez Gene ID
78038
Gene Name
methylcrotonoyl-Coenzyme A carboxylase 2 (beta)
Gene Aliases
4930552N12Rik, MCCB
Chromosome Location
Chr.13: 99948532 - 100015639 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030026.2 NP_084302.1 8 - 9 927 76
GenBank mRNA AK019760.1 - 2 - 3 108 76
AK132265.1 - 8 - 9 907 76
AK143233.1 - 8 - 9 928 76
AK145564.1 - 8 - 9 879 76
AK146844.1 - 8 - 9 887 76
AK146865.1 - 8 - 9 877 76
BC141193.1 - 8 - 9 893 76
BC141194.1 - 8 - 9 893 76

View Full Product Details