Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pde8b
Entrez Gene ID
218461
Gene Name
phosphodiesterase 8B
Gene Aliases
B230331L10Rik, C030047E14Rik
Chromosome Location
Chr.13: 95024105 - 95250397 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170669.1 NP_001164140.1 15 - 16 2076 90
NM_172263.2 NP_758467.2 16 - 17 1809 90
GenBank mRNA AK043680.1 - 15 - 16 2069 90
AK045991.1 - 15 - 16 2078 90
AK083166.1 - 15 - 16 1714 90
AK138301.1 - 5 - 6 743 90
AK154105.1 - 1 - 2 99 90
AK170192.1 - 1 - 2 88 90
BC032286.1 - 16 - 17 1821 90
EF644173.1 - 18 - 19 1821 90

View Full Product Details