Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nr5a2
Entrez Gene ID
26424
Gene Name
nuclear receptor subfamily 5, group A, member 2
Gene Aliases
AU020803, D1Ertd308e, Ftf, LRH-1, UF2-H3B
Chromosome Location
Chr.1: 136842562 - 136960448 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159769.2 NP_001153241.1 4 - 5 455 108
NM_030676.3 NP_109601.1 4 - 5 540 108
GenBank mRNA AB221629.1 - 4 - 5 382 108
AK054487.1 - 2 - 3 109
BB652042.1 - 4 - 5 426 108
BC137845.1 - 4 - 5 419 108
DQ133942.1 - 4 - 5 455 108
M81385.1 - 4 - 5 540 108

View Full Product Details