Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmco1
Entrez Gene ID
68944
Gene Name
transmembrane and coiled-coil domains 1
Gene Aliases
1190006A08Rik, 4930403O06Rik, AA109065, AU022572, ESTM39
Chromosome Location
Chr.1: 167308670 - 167333978 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039483.1 NP_001034572.1 2 - 3 346 81
GenBank mRNA AK004504.1 - 2 - 3 188 81
AK015068.1 - 2 - 3 637 81
AK037709.1 - 2 - 3 281 81
AK162344.1 - 2 - 3 285 81
BC011457.1 - 2 - 3 220 81
BC019761.1 - 2 - 3 264 81
BC020098.1 - 2 - 3 276 81

View Full Product Details