The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252494.1 | NP_001239423.1 | 14 - 15 | 1932 | 94 |
NM_001252496.1 | NP_001239425.1 | 14 - 15 | 1932 | 94 | |
NM_001252497.1 | NP_001239426.1 | 9 - 10 | 2190 | 94 | |
NM_175258.4 | NP_780467.2 | 14 - 15 | 1932 | 94 | |
GenBank mRNA | AK045540.1 | - | 14 - 15 | 1874 | 94 |
AK143331.1 | - | 2 - 3 | 319 | 94 | |
AK164343.1 | - | 9 - 10 | 2163 | 94 | |
AK167055.1 | - | 14 - 15 | 1933 | 94 | |
AK220459.1 | - | 3 - 4 | 335 | 94 | |
BC141182.1 | - | 14 - 15 | 1831 | 94 | |
BC145503.1 | - | 14 - 15 | 1831 | 94 |