Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rapgef6
Entrez Gene ID
192786
Gene Name
Rap guanine nucleotide exchange factor (GEF) 6
Gene Aliases
A530068K01, AI844632, BB085664, C030018K18Rik, PDZ-GEF2, Pdzgef2, RA-GEF-2, Ragef2, mKIAA4052
Chromosome Location
Chr.11: 54521754 - 54699286 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252494.1 NP_001239423.1 21 - 22 3292 106
NM_001252496.1 NP_001239425.1 21 - 22 3292 106
NM_175258.4 NP_780467.2 20 - 21 3277 106
GenBank mRNA AK143331.1 - 8 - 9 1664 106
AK220459.1 - 9 - 10 1680 106
BC025553.1 - 3 - 4 771 106
BC092525.1 - 3 - 4 844 106
BC141182.1 - 21 - 22 3191 106
BC145503.1 - 21 - 22 3191 106

View Full Product Details