The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159396.1 | NP_001152868.1 | 1 - 2 | 219 | 70 |
GenBank mRNA | BC003821.1 | - | 1 - 2 | 197 | 70 |
BY328342.1 | - | 1 - 2 | 222 | 70 |