Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mfsd4
Entrez Gene ID
213006
Gene Name
major facilitator superfamily domain containing 4
Gene Aliases
A230072B04, A930031D07Rik, AI850289
Chromosome Location
Chr.1: 132022806 - 132068062 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114662.1 NP_001108134.1 9 - 10 1649 110
GenBank mRNA AK085484.1 - 6 - 7 1065 110

View Full Product Details