Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bbs7
Entrez Gene ID
71492
Gene Name
Bardet-Biedl syndrome 7 (human)
Gene Aliases
8430406N16Rik
Chromosome Location
Chr.3: 36573140 - 36613489 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027810.3 NP_082086.2 11 - 12 1335 79
GenBank mRNA AF521645.1 - 11 - 12 1334 79
AK050291.1 - 11 - 12 1333 79
AK132661.1 - 11 - 12 1263 79
BC031505.1 - 11 - 12 1285 79

View Full Product Details