Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Egfem1
Entrez Gene ID
75740
Gene Name
EGF-like and EMI domain containing 1
Gene Aliases
6130401L20Rik, Efcab11
Chromosome Location
Chr.3: 29080745 - 29691058 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167748.1 NP_001161220.1 1 - 2 359 58
NM_029412.1 NP_083688.1 1 - 2 359 58
GenBank mRNA AK018073.1 - 1 - 2 362 58
AK032017.1 - 1 - 2 203 58
BC117810.1 - 1 - 2 165 58

View Full Product Details