The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029412.1 | NP_083688.1 | 6 - 7 | 883 | 68 |
GenBank mRNA | AK032017.1 | - | 6 - 7 | 727 | 68 |