The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286865.1 | NP_001273794.1 | 1 - 2 | 219 | 97 |
NM_011275.3 | NP_035405.2 | 1 - 2 | 219 | 97 | |
NR_104608.1 | 1 - 2 | 219 | 97 | ||
GenBank mRNA | AF048993.1 | - | 1 - 2 | 180 | 97 |
AK011680.1 | - | 1 - 2 | 192 | 97 | |
BC019411.1 | - | 1 - 2 | 148 | 97 | |
BY274268.1 | - | 1 - 2 | 220 | 97 | |
CA554498.1 | - | 1 - 2 | 170 | 97 | |
CJ073105.1 | - | 1 - 2 | 195 | 97 |