Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Cd48
- Entrez Gene ID
-
12506
- Gene Name
- CD48 antigen
- Gene Aliases
-
AI449234, AW610730, BCM1, BLAST, BLAST-1, BLAST1, Bcm-1, MEM-102, SLAMF2, Sgp-60
- Chromosome Location
-
Chr.1: 171681421 - 171705257 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK088027.1 |
- |
2 - 3 |
463 |
90 |
View Full Product Details