The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243062.1 | NP_001229991.1 | 7 - 8 | 1049 | 80 |
GenBank mRNA | AF009328.1 | - | 7 - 8 | 1083 | 80 |
BC025154.1 | - | - | 580 | 80 |