Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Fcgr2b
- Entrez Gene ID
-
14130
- Gene Name
- Fc receptor, IgG, low affinity IIb
- Gene Aliases
-
AI528646, CD32, F630109E10Rik, Fc[g]RII, FcgRII, Fcgr2, Fcgr2a, Fcr-2, Fcr-3, Ly-17, Ly-m20, LyM-1, Lym-1, fcRII
- Chromosome Location
-
Chr.1: 170960558 - 170976071 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
M16367.1 |
- |
1 - 2 |
115 |
98 |
View Full Product Details