Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mettl18
Entrez Gene ID
69962
Gene Name
methyltransferase like 18
Gene Aliases
2810422O20Rik, C78571
Chromosome Location
Chr.1: 163994891 - 163997243 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK139786.1 - 1 - 2 188 68
BC052693.1 - 1 - 2 162 68

View Full Product Details