Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Scyl3
Entrez Gene ID
240880
Gene Name
SCY1-like 3 (S. cerevisiae)
Gene Aliases
1200016D23Rik, 6030457O16, AW214499, Pace1
Chromosome Location
Chr.1: 163927958 - 163955126 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286002.1 NP_001272931.1 7 - 8 1016 132
NM_001286003.1 NP_001272932.1 7 - 8 1228 132
NM_028776.5 NP_083052.1 7 - 8 1203 132
GenBank mRNA AK004809.2 - 7 - 8 943 132
AK030824.1 - 7 - 8 1178 132
AK044100.1 - 7 - 8 1189 132
AK162937.1 - 7 - 8 1017 132
AK184387.1 - 1 - 2 96 132
BC043085.1 - 7 - 8 904 132
BC066800.1 - 7 - 8 888 132

View Full Product Details