Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Scyl3
Entrez Gene ID
240880
Gene Name
SCY1-like 3 (S. cerevisiae)
Gene Aliases
1200016D23Rik, 6030457O16, AW214499, Pace1
Chromosome Location
Chr.1: 163927958 - 163955126 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286002.1 NP_001272931.1 8 - 9 1106 143
NM_001286003.1 NP_001272932.1 8 - 9 1318 143
NM_028776.5 NP_083052.1 8 - 9 1293 143
GenBank mRNA AK004809.2 - 8 - 9 1033 143
AK030824.1 - 8 - 9 1268 143
AK044100.1 - 8 - 9 1279 143
AK162937.1 - 8 - 9 1107 143
AK184387.1 - 2 - 3 186 143
BC043085.1 - 8 - 9 994 143
BC066800.1 - 8 - 9 978 143

View Full Product Details