The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083897.1 | NP_001077366.1 | 5 - 6 | 864 | 56 |
GenBank mRNA | AK090278.1 | - | 5 - 6 | 850 | 56 |
AK169385.1 | - | 5 - 6 | 854 | 56 | |
AK202400.1 | - | 2 - 3 | 125 | 56 | |
AY764247.1 | - | 5 - 6 | 870 | 56 |