The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037170.2 | NP_001032247.1 | 3 - 4 | 460 | 70 |
GenBank mRNA | AK012264.1 | - | 3 - 4 | 390 | 70 |
AK028388.1 | - | 3 - 4 | 461 | 70 | |
BC058978.1 | - | 2 - 3 | 351 | 70 |