Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Frem2
Entrez Gene ID
242022
Gene Name
Fras1 related extracellular matrix protein 2
Gene Aliases
6030440P17Rik, 8430406N05Rik, Gm409, b2b1562Clo, my, ne, nv1
Chromosome Location
Chr.3: 53513938 - 53657912 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172862.3 NP_766450.2 8 - 9 6622 74
GenBank mRNA AB236662.1 - 8 - 9 6622 74
AJ833643.1 - 8 - 9 6351 74
CN530725.1 - 3 - 4 508 74

View Full Product Details