Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Pogk
- Entrez Gene ID
-
71592
- Gene Name
- pogo transposable element with KRAB domain
- Gene Aliases
-
9130401E23Rik, BASS2, mKIAA1513
- Chromosome Location
-
Chr.1: 166393610 - 166409858 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK048752.1 |
- |
2 - 3 |
210 |
84 |
View Full Product Details