Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Chit1
Entrez Gene ID
71884
Gene Name
chitinase 1 (chitotriosidase)
Gene Aliases
2300002L19Rik
Chromosome Location
Chr.1: 134111242 - 134151541 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284524.1 NP_001271453.1 5 - 6 546 70
NM_001284525.1 NP_001271454.1 5 - 6 582 70
NM_027979.2 NP_082255.1 5 - 6 546 70
GenBank mRNA AK009012.1 - 5 - 6 547 70
AY458654.1 - 5 - 6 546 70
AY536287.1 - 4 - 5 420 70
BC138765.1 - 5 - 6 548 70

View Full Product Details