Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Chit1
Entrez Gene ID
71884
Gene Name
chitinase 1 (chitotriosidase)
Gene Aliases
2300002L19Rik
Chromosome Location
Chr.1: 134111242 - 134151541 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284524.1 NP_001271453.1 7 - 8 836 62
NM_001284525.1 NP_001271454.1 7 - 8 872 62
NM_027979.2 NP_082255.1 7 - 8 836 62
GenBank mRNA AK009012.1 - 7 - 8 837 62
AY458654.1 - 7 - 8 836 62
AY536287.1 - 6 - 7 710 62
BC138765.1 - 7 - 8 838 62

View Full Product Details