Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Chit1
Entrez Gene ID
71884
Gene Name
chitinase 1 (chitotriosidase)
Gene Aliases
2300002L19Rik
Chromosome Location
Chr.1: 134111242 - 134151541 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284525.1 NP_001271454.1 10 - 11 1287 64
NM_027979.2 NP_082255.1 10 - 11 1251 64
GenBank mRNA AY458654.1 - 10 - 11 1251 64
AY536287.1 - 9 - 10 1125 64
BC138765.1 - 10 - 11 1253 64

View Full Product Details